Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NNMTi

Nicotinamide N-methyltransferase (NNMT) inhibitor; promotes myoblast differentiation

$104.00 - $440.00

Specifications

CAS 42464-96-0
Chemical Name or Material 5-Amino-1-methylquinolinium iodide
Molecular Formula C10H11IN2
Purity 97%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
69-001-0
View Documents
Tocris Bioscience™
6900/10
10 mg
Each for $104.00
Add to cart
 
69-005-0
View Documents
Tocris Bioscience™
6900/50
50 mg
Each for $440.00
Add to cart
 
Description

Description

  • Nicotinamide N-methyltransferase (NNMT) inhibitor (IC50 = 1.2 μM).
  • Promotes myoblast differentiation in C2C12 cells.
  • Activates senescent muscle stem cells (muSCs) and improves regenerative capacity of aged skeletal muscle.
  • Enhances proliferation, fusion and regenerative capacity of muSCs in aged mice.
Specifications

Specifications

42464-96-0
5-Amino-1-methylquinolinium iodide
C10H11IN2
97%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit