Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 189841AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-2304


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The antibody can recognize isoforms 1, 2, and 4 of NOS1.

NOS activity requires homodimerization as well as three cosubstrates (L-arginine, NADPH and O2) and five cofactors or prosthetic groups (FAD, FMN, calmodulin, tetrahydrobiopterin and heme). Several distinct NOS isoforms have been described and been shown to represent the products of three distinct genes. These include two constitutive Ca2+/CaM-dependent forms of NOS, including NOS1 (also designated ncNOS) whose activity was first identified in neurons, and NOS3 (also designated ecNOS), first identified in endothelial cells.
Specifications

Specifications

nNOS
Polyclonal
Unconjugated
NOS1
bNOS, Constitutive NOS, IHPS1, N NOS, NC NOS, Neuronal NOS, Nitric oxide synthase, brain, nNOS, NOS, NOS type I, NOS1
Rabbit
Antigen Affinity Chromatography
RUO
18125, 4842
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P29475, Q9Z0J4
Nos1
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.