Learn More
Ace Glass, Inc. 19mm ID x 30 cm chromatography column, #3 stopper top, 250mL reservoir, porosity B filter disc
SDP
Supplier:  Ace Glass, Inc. 590737
Encompass_Preferred
19mm I.D. x 30cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper. Includes porosity B filter disc.
Catalog No. 50-972-592