Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Noble Agar, Ultrapure

Gelling agent essentially free of impurities. Thermo Scientific™ Noble Agar (Ultrapure) is mainly used in biochemistry and molecular biology procedures where increased purity is required.

Manufacturer:  Thermo Scientific™ J1090722

 View more versions of this product

Catalog No. AAJ1090722


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

  • Format: Pale cream powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288
Typical Agar Preparation
Plates: 15g/L of media; 30 to 35mL per 90mm plate
Top Agar: 7g/L of media
Autoclave for 20 minutes and swirl gently to evenly distribute the agar. If antibiotics are to be added, allow the agar to cool to 50°C. When pouring plates, gently swirl the medium. A Bunsen burner flame can be passed over the surface of the plates to remove any air bubbles. After the medium has hardened, invert the plates and store at 4°C
Specifications

Specifications

Powder
Plastic Bottle
Ultrapure
Nobel Agar
100g
Pale cream
Powder
Used as a gelling agent in biochemistry and molecular biology procedures where increased purity is required.
Safety and Handling

Safety and Handling

EINECSNumber : 232-658-1

Recommended Storage : Ambient temperatures

Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit