Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOC3L Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen NOC3L
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP156497
SDP
View Documents
Novus Biologicals
NBP156497
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

NOC3L Polyclonal specifically detects NOC3L in Human samples. It is validated for Western Blot.
Specifications

Specifications

NOC3L
Polyclonal
Rabbit
Protein Kinase
AD24NOC3-like protein, C10orf117, chromosome 10 open reading frame 117, Factor for adipocyte differentiation 24, FAD24NOC3 protein homolog, FLJ12820, nucleolar complex associated 3 homolog (S. cerevisiae), nucleolar complex protein 3 homolog, Nucleolar complex-associated protein 3-like protein
NOC3L
IgG
Western Blot
Unconjugated
RUO
Q8WTT2
64318
Synthetic peptides corresponding to NOC3L(nucleolar complex associated 3 homolog (S. cerevisiae)) The peptide sequence was selected from the N terminal of NOC3L. Peptide sequence TLKKYRKEQRKLRQAVKDAVSKKPIPLENPKEKRPGKRIEREEEEEEEAL.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.