Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Noggin Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040413AF350
Description
Noggin Polyclonal antibody specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Noggin | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Alexa Fluor 350 | |
noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1 | |
Synthetic peptide comprising residues 156-170 [PVLYAWNDLGSRFWP] of the human Noggin protein | |
0.1 mL | |
Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
9241 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction