Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Noggin Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: OTI1H8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273030UV
Description
Noggin Monoclonal specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Noggin | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NOG | |
Recombinant protein fragment corresponding to amino acids 28-232 of human Noggin (NP_005441) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1H8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1 | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
9241.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title