Learn More
Thermo Scientific™ Noggin, Mouse
Supplier:  Thermo Scientific™ J67203YAM
Catalog No. AAJ67203YAM
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μg
-30°C to -10°C
Recombinant Murine NOG; NOG, mouse
Lyophilized Powder
White
46.4 kDa
Noggin, Mouse
Product Suggestions
Product Suggestions