Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nogo Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 16 publications
$206.50 - $530.25
Specifications
| Antigen | Nogo |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Simple Western 1:1000, ELISA reported in scientific literature (PMID 17413036), Immunohistochemistry reported in scientific literature (PMID 30208932), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27132996), Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056681T
![]() |
Novus Biologicals
NB10056681SS |
0.05 mL |
Each for $206.50
|
|
|||||
NB10056681
![]() |
Novus Biologicals
NB10056681 |
0.2 mL |
Each for $530.25
|
|
|||||
Description
Nogo Polyclonal specifically detects Nogo in Human, Mouse, Rat, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Nogo | |
| Western Blot 1:500-1:2000, Simple Western 1:1000, ELISA reported in scientific literature (PMID 17413036), Immunohistochemistry reported in scientific literature (PMID 30208932), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27132996), Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Sheep | |
| Q9NQC3 | |
| 57142 | |
| A synthetic peptide to an internal portion of human Nogo A (between amino acids 1-50) [UniProt Q9NQC3]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS with 0.05% Sodium Azide | |
| ASY, foocen, Human NogoA, KIAA0886, My043 protein, Nbla00271, Nbla10545, neurite growth inhibitor 220, Neurite outgrowth inhibitor, Neuroendocrine-specific protein, Neuroendocrine-specific protein C homolog, NI220/250, Nogo protein, NOGO-A, Nogo-B, Nogo-C, NOGOC, NOGORTN4-B1, NSP, NSP-CL, reticulon 4, reticulon 5, reticulon-4, reticulon-5, RTN4-A, RTN4-B2, RTN4-C, RTN-x | |
| RTN4 | |
| IgG | |
| Protein G purified | |
| This recognizes Nogo-A (approx. 220 kD)and Nogo-B2 and B1 (approx. 50 kD and 48 kD in human and 43 kD and 41 kD in mouse and rat, respectively). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title