Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nogo Receptor/NgR Rat anti-Mouse, HRP, Clone: RM0126-3D66, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122503H
Description
Nogo Receptor/NgR Monoclonal antibody specifically detects Nogo Receptor/NgR in Mouse samples. It is validated for Western Blot.Specifications
Nogo Receptor/NgR | |
RM0126-3D66 | |
Western Blot | |
Purified | |
RTN4R | |
Mouse recombinant protein of the extracellular domain of Nogo Receptor | |
Protein G purified | |
Store at 4C in the dark. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of Nogo Receptor. |
Western Blot | |
HRP | |
PBS with No Preservative | |
NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor | |
Rat | |
IgG2 | |
0.1 ml | |
Primary | |
65078 | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title