Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOL1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30926125UL

 View more versions of this product

Catalog No. NB123424


Add to cart

Description

Description

NOL1 Polyclonal specifically detects NOL1 in Rat samples. It is validated for Western Blot.
Specifications

Specifications

NOL1
Unconjugated
PBS buffer, 2% sucrose
EC 2.1.1, EC 2.1.1.-, MGC149287, MGC149288, NOL1/NOP2/Sun domain family, member 1, NOL1MGC117384, NOP120, NOP2 nucleolar protein homolog (yeast), NOP2/Sun domain family, member 1, NSUN1, Nucleolar protein 1, nucleolar protein 1 (120kD), nucleolar protein 1, 120kDa, Nucleolar protein 2 homolog, nucleolar protein 2 homolog (yeast), p120, Proliferating-cell nucleolar antigen p120, Proliferation-associated nucleolar protein p120, putative ribosomal RNA methyltransferase NOP2
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat NOL1 (NP_001178714). Peptide sequence TKKEKRGPGRKARKQKGAETELIRFLPAAGDENFKRLSSRARKRAAKRRS
25 μg
Core ESC Like Genes, Stem Cell Markers
Primary
4839
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit