Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOL10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560229
Description
Immunogen sequence: AENNVCDINS VHGLFATGTI EGRVECWDPR TRNRVGLLDC ALNSVTADSE INSLPTISAL KFNGALTMAV GTTTGQ Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
NOL10 is a protein coding gene.
Specifications
NOL10 | |
Polyclonal | |
Unconjugated | |
NOL10 | |
Gm67; H_NH0074G24.1; NOL10; Nucleolar protein 10; polyglutamine binding protein 5; PQBP5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79954 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BSC4 | |
NOL10 | |
Recombinant protein corresponding to Human NOL10. Recombinant protein control fragment (Product #RP-96029). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction