Learn More
Aviva Systems Biology Corp NOL3 Antibody - middle region (AVARP13001_P050) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP13001_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NOL3. Alternate names: ARC, MYC, NOP, NOP30, MYPApplicationWB. Species reactivity: Human
Catalog No. 50-014-62597