Learn More
NOL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 163611AP
Catalog No. 50-173-0893
Add to cart
Description
Description
The nucleolus is a dense subnuclear membraneless organelle that assembles around clusters of rRNA genes and functions in ribosome biogenesis. This gene encodes a nucleolar RNA-associated protein that is highly conserved between species. RNase treatment of permeabilized cells indicates that the nucleolar localization is RNA dependent. Further studies suggest that the protein is associated with ribosome biogenesis through an interaction with pre-rRNA primary transcripts. Alternative splicing has been observed at this locus and two splice variants encoding distinct isoforms have been identified.
Specifications
Specifications
NOL6
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R5K4, Q9H6R4
NOL6, NRAP, Nucleolar protein 6, UTP22
Rabbit
IgG
150 μL
-20°C
Primary
230082, 313167, 65083
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
NOL6
Liquid
NOL6
NOL6 Fusion Protein Ag8368
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat