Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOM1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP276357

Catalog No. NBP276357


Add to cart

Description

Description

NOM1 Polyclonal antibody specifically detects NOM1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

NOM1
Unconjugated
0.09% Sodium Azide
C7orf3, chromosome 7 open reading frame 3, FLJ16401, nucleolar MIF4G domain-containing protein 1, nucleolar protein with MIF4G domain 1, SGD1, SGD1 homolog
Rabbit
IgG
100 μL
Primary
64434.0
Immunoprecipitation, Western Blot
Western Blot 1:1000, Immunoprecipitation 50-100 μL/mg lysate
Purified
NOM1
Immunogen sequence between amino acids 200 and 250
Antigen Affinity-purified
Store at 2-8°C/ 1 year from date of receipt
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit