Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nomega-Monomethyl-L-arginine Acetate 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
53308-83-1
C9H20N4O4
248.283
MFCD00069311
IKPNWIGTWUZCKM-JEDNCBNOSA-N
tilarginine acetate,l-nmma,l-nmma acetate,ng-monomethyl-l-arginine acetate,nomega-me-l-arg,unii-2fl3530af2,ng-me-l-arg, acoh,tilarginine acetate usan,ng-methyl-l-arginine acetate salt,l-ornithine, n5-imino methylamino methyl-cas
135242
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
53308-83-1
C9H20N4O4
248.283
MFCD00069311
IKPNWIGTWUZCKM-JEDNCBNOSA-N
tilarginine acetate,l-nmma,l-nmma acetate,ng-monomethyl-l-arginine acetate,nomega-me-l-arg,unii-2fl3530af2,ng-me-l-arg, acoh,tilarginine acetate usan,ng-methyl-l-arginine acetate salt,l-ornithine, n5-imino methylamino methyl-cas
135242
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
Quantity 100mg
Color White
Formula Weight 248.28
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Nomega-Monomethyl-L-arginine Acetate

RTECSNumber : RM2982000

TSCA : No