Learn More
Mopec Non-Elevating Rotating Autopsy Table CE Series Available on GSA/VA Contract for Federal Government customers only.
Pedestal style table that incorporates a rotating mechanism within the pedestal. Mopec™ Non-Elevating Rotating Autopsy Table CE Series has a downdraft ventilation system which helps ventilate the body during an autopsy.
Manufacturer:  Mopec CE300
Catalog No. 22-444-026
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • All stainless steel construction
  • 14 gauge table top, sink and grid plates
  • 11 gauge pedestal
  • All type 304 stainless steel with #4 finish
  • 180 degrees of total table rotation
  • Large sink integral to table top
  • Hydro aspirators designed with reversing flow valve
  • Includes hose hanger and 8 foot clear PVC tubing
  • Cold water control valve with vacuum brake swing spout
  • Single lever hot/cold water fixture with swing spout and vacuum breaker
  • 4 interchangeable removable grid plates, stainless steel construction with 0.5 in. recess and 0.375 in. diameter holes on 2 in. square center
  • Heavy duty brass chrome plated hand piece
  • Concealed pressure control/shut off
  • Access panel
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
1 Year Warranty
Specifications
Specifications
2 GFCI duplex outlet with waterproof cover
Down draft
30 in.
76.2cm
100 in.
Autopsy Tables
37 in.
93.98cm
Holds up to 750lbs
254cm