Learn More
Americanbio IncSupplier Diversity Partner NON-FAT DRY MILK OMNIBLOK100GR DFS Item
Manufacturer:  Americanbio IncSupplier Diversity Partner AB1010900100
This item has a minimum order quantity of 6 per supplier requirements. Omniblok non-fat dry milk is exceptionally pure and is specifically tested As a blocking agent for use during western blotting To block non-specific antibody binding and to reduce background signal. Typically, western blotting and elisa methods call for the addition of a blocking agent Prior to adding a primary antibody Usa origin only
Catalog No. NC0166631