Learn More
Steritool NON-MAG SSTEEL MALLET
Supplier:  Steritool 10190/40190
Encompass_Preferred
Non-Magnetic Stainless Steel Mallet. SS Handle. Nylon Head - 316 Stainless Steel & Nylon.
Catalog No. NC1705091