Alfa Aesar™ Non Metals, plasma standard solution, Specpure™, As, B, P, S, Se, Si, Te at 100µg/mL

Manufacturer: Alfa Aesar™ 44267AE

Quantity 100mL

 View more versions of this product

Catalog No. AA44267AE

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00144997
    UN Number UN3264
    Chemical Name or Material Non Metals plasma standard solution, As, B, P, S, Se, Si, Te at 100μg/mL
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00144997
    UN Number UN3264
    Health Hazard 1 Danger
    Quantity 100mL
    Avg. Mol. Wt. or Mol. Wt. Range Matrix: 5% HCl/tr. F-
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Physical Form Liquid
    Chemical Name or Material Non Metals plasma standard solution, As, B, P, S, Se, Si, Te at 100μg/mL
    Grade Specpure™

    GHS H Statement
    H301-H311-H314-H318-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if inhaled.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit