Learn More
MilliporeSigma™ Supelco™ Non-Polar Fused Silica Tubing
Ideal for use with injection solvents such as alkanes, carbon disulfide, and ethers
Supplier: MilliporeSigma™ Supelco™ 25723
Description
- Fused silica tubing refers to tubing with a protective coating
- Use as transfer lines, guard columns, or retention gaps
- Can be used to make customized columns
- Ideal for use with injection solvents such as alkanes, carbon disulfide, and ethers
- Composition: Methyl
- Deactivated
- Non-polar
Specifications
Injection Solvents Such as Alkanes, Carbon Disulfide, and Ethers | |
Gas Chromatography (GC) | |
3 m |
Tubing | |
0.32 mm | |
Silica |