Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Non-Reflective Hi-Vis Safety Vests

Pyramex™ Non-Reflective Hi-Vis Safety Vests do not meet ASNI/CSA safety standards

$3.90

Specifications

Closure Type 5 in. Hook and Loop
Material Polyester Mesh
Size Universal
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19-555-442
View Documents
Pyramex™
RV110NS
Hi-Vis Lime
Each for $3.90
Case of 12 EA for $46.80
Add to cart
 
19-555-444
View Documents
Pyramex™
RV120NS
Hi-Vis Orange
Each for $3.90
Case of 12 EA for $46.80
Add to cart
 
Description

Description

  • Front: 5 in. Hook & Loop closure
  • Sides: 0.75 x 7 in. elastic bands
Specifications

Specifications

5 in. Hook and Loop
Polyester Mesh
Universal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit