Learn More
Novus Biologicals™ Non-species specific 11-dehydro-TXB2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP262129
 View more versions of this product
Catalog No. NBP262129
Add to cart
Description
Description
The Non-species specific 11-dehydro-TXB2 ELISA Kit (Colorimetric) measures non-species specific 11-dehydro-TXB2 in culture supernatants, urine, serum.
5 Kits
Specifications
Specifications
9.8 - 10,000 pg/ml
ELISA
ELISA Kit (Colorimetric)
Culture Supernatant,Serum,Urine
11-dehydro-TXB2
4.31 pg/ml
Non-species specific
5 Kit
Storage of components varies. See protocol for specific instructions.