Learn More
Novus Biologicals™ Non-species specific 8-Isoprostaglandin F2a ELISA Kit (Colorimetric)
Non-species specific 8-Isoprostaglandin F2a ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP2621415KIT
Catalog No. N2621415KIT
Add to cart
Description
Description
The Non-species specific 8-Isoprostaglandin F2a ELISA Kit (Colorimetric) measures non-species specific 8-Isoprostaglandin F2a in plasma, serum, tissue, cell lysate, urine.
Specifications
Specifications
Cell Lysates, Plasma, Serum, Tissue, Urine
160 to 100,000 pg/ml
40.0 pg/ml
Non-species specific
5 Kit
ELISA Kit (Colorimetric)
Store at 4°C