Learn More
Novus Biologicals™ Non-species specific 8-Isoprostaglandin F2a ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP262132
Description
The Non-species specific 8-Isoprostaglandin F2a ELISA Kit (Colorimetric) measures non-species specific 8-Isoprostaglandin F2a in culture supernatants, tissue, urine, plasma.Specifications
6.1 - 100,000 pg/ml | |
ELISA | |
ELISA Kit (Colorimetric) | |
Culture Supernatant,Plasma,Tissue,Urine | |
8-Isoprostaglandin F2a |
16.3 pg/ml | |
Non-species specific | |
1 Kit | |
Store at 4°C |