Learn More
Novus Biologicals™ Angiotensin II ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP262135
Catalog No. NBP262135
Add to Cart
Description
Description
The Non-species specific Angiotensin II ELISA Kit (Colorimetric) measures non-species specific Angiotensin II in plasma,serum.
Specifications
Specifications
3.9-10,000 pg/ml
Crossreactivity: Angiotensin II (100%), Angiotensin A (100%), Angiotensin III (100%), Angiotensin IV (100%), Angiotensin I (0.319%), Angiotensin (1-9) (0.103%), LVV-hemorphin 7 (0.067%), Angiotensin (1-7) (0.053%), Bradykinin (0.022%)
Plasma, Serum
183
Non-species specific
Storage of components varies. See protocol for specific instructions.
4.6 pg/ml
ELISA Kit (Colorimetric)
ELISA
AGT
1 Kit
Angiotensin II
Recommendations
Recommendations