Learn More
Novus Biologicals Non-species specific Dibromo-tyrosine ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP26192896TESTS
Description
The Non-species specific Dibromo-tyrosine ELISA Kit (Colorimetric) measures non-species specific Dibromo-tyrosine in plasma, urine.96 Tests
Specifications
ELISA Kit (Colorimetric) | |
Intra-Assay Precision: Three samples of known concentration were assayed thirty times on one plate; the intra-assay coefficient of variation of the Dibromo-tyrosine ELISA has been determined to be < 10%. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Non-species specific | |
Plasma, Urine |
0.078 - 5 ug/mL | |
ELISA | |
Dibromo-tyrosine | |
96 Tests | |
0.04 ug/ml |