Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Non-species specific LVV Hemorphin 7 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP262158
Description
The Non-species specific LVV Hemorphin 7 ELISA Kit (Colorimetric) measures non-species specific LVV Hemorphin 7 in serum, tissue homogenate.Specifications
ELISA Kit (Colorimetric) | |
Crossreactivity: LVV Hemorphin 7 (100%) and <0.003%: Leu-Valorphin-Arg (LVV Hemorphin-6), Valorphin, Angiotensin (1-12), Angiotensin I, Angiotensin (1-9), Angiotensin II, Angiotensin (1-7), Angiotensin A, Angiotensin III, Angiotensin IV | |
Storage of components varies. See protocol for specific instructions. | |
Non-species specific | |
Serum, Tissue Homogenates |
9.8 - 10,000 pg/ml | |
ELISA | |
LVV Hemorphin 7 | |
1 Kit | |
6.1 pg/ml |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title