Learn More
Novus Biologicals Non-species specific Melatonin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP262160
Description
The Non-species specific Melatonin ELISA Kit (Colorimetric) measures non-species specific Melatonin in saliva, serum, plasma, fruit samples.Specifications
ELISA Kit (Colorimetric) | |
ELISA | |
Melatonin | |
1 Kit | |
0.162 ng/ml |
0.08-50 ng/ml | |
Storage of components varies. See protocol for specific instructions. | |
Non-species specific | |
Fruit, Plasma, Saliva, Serum |