Learn More
Novus Biologicals™ Non-species specific Prostacyclin ELISA Kit (Colorimetric)
Non-species specific Prostacyclin ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP2621715KIT
Catalog No. N2621715KIT
Add to cart
Description
Description
The Non-species specific Prostacyclin ELISA Kit (Colorimetric) measures non-species specific Prostacyclin in culture supernatants, urine.
Specifications
Specifications
Culture Supernatant, Urine
7.81 to 2,000 pg/ml
6.58 pg/ml
Non-species specific
5 Kit
ELISA Kit (Colorimetric)
Store at 4°C