Learn More
Novus Biologicals Non-species specific Prostacyclin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP262171
Description
The Non-species specific Prostacyclin ELISA Kit (Colorimetric) measures non-species specific Prostacyclin in culture supernatants, urine.Specifications
ELISA Kit (Colorimetric) | |
ELISA | |
Prostacyclin | |
1 Kit | |
6.58 pg/ml |
7.81 - 2,000 pg/ml | |
Store at 4°C | |
Non-species specific | |
Culture Supernatant, Urine |