Learn More
Henke-Sass, Wolf Non-Sterile 2-Part Syringes
These syringes feature a 2-part construction containing no rubber and eliminating the need for silicone oil. Made from polypropylene and polyethylene.
Supplier: Henke-Sass, Wolf ABC3LL
Description
- Disposable Syringes
- 2-Part Syringe
- Unsterile
- Polypropylene Barrel
- Polyethylene Plunger
- Single-use
- Latex Free
- ISO
- Chromatography
- Laboratory Use
Specifications
Not Autoclavable | |
Polyethylene (Plunger), Polypropylene (Barrel) | |
100 Syringes | |
Yes | |
3 mL | |
Yes |
ISO 13485 | |
Clear | |
Yes | |
Bulk | |
Luer Lock | |
Non-sterile |