Learn More
Honeywell™ North™ Non-Vented Short Brim Hard Hat
Cost-effective certified head protection solution offers enhanced performance, all-day comfort and durability
Manufacturer:  Honeywell™ North™ NSB10002
 View more versions of this product
Catalog No. 19-185-001
Add to cart
Description
Description
  • High-density polyethylene (HDPE)
  • Four-point ratchet suspension
  • Dual-locking mechanism
  • Men's
Specifications
Specifications
No
ANSI (ANSI Z89.1 – 2014) standard, CE/EN (EN 397:2012 +AI:2012, EN 50365:2002 (non-vented versions) standard, CSA (CSA Z94.1-15) standard, NOM standard
Compatible with CP5005 plastic face-shield bracket, 3796VS1 helmet adapter for cap mounted earmuffs
HDPE
Universal
Short Brim
Yellow
Construction, Mining, Manufacturing, Oil and gas, Ship building, Traffic safety, Utilities
Type 1