Alfa Aesar™ Nonacarbonyldiiron, 99%

Manufacturer: Alfa Aesar™ 1306218

Quantity 50g

 View more versions of this product

Catalog No. AA1306218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15321-51-4
    Linear Formula Fe2(CO)9
    Formula Weight 363.79
    MDL Number MFCD00151465
    UN Number UN3179
    Synonym Diiron nonacarbonyl; Iron nonacarbonyl
    Chemical Name or Material Nonacarbonyldiiron

    Specifications

    Formula Weight 363.79
    Health Hazard 3 GHS P Statement
    P210-P280f-P304+P340-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Immediately call a POISON CENTER or doctor/physician.
    Melting Point ≈100°C (dec.)
    Sensitivity Air Sensitive
    Health Hazard 2 GHS H Statement
    H228-H301-H330
    Flammable solid.
    Toxic if swallowed.
    Fatal if inhaled.
    TSCA TSCA
    Physical Form Crystalline
    CAS 15321-51-4
    UN Number UN3179
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
    Density 2.85g/mL
    Quantity 50g
    MDL Number MFCD00151465
    Synonym Diiron nonacarbonyl; Iron nonacarbonyl
    Color Orange
    Linear Formula Fe2(CO)9
    Chemical Name or Material Nonacarbonyldiiron
    Assay 99%

    GHS H Statement
    H228-H301-H330
    Flammable solid.
    Toxic if swallowed.
    Fatal if inhaled.

    GHS P Statement
    P210-P280f-P304+P340-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit