Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonadecanoic acid, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
646-30-0
C19H38O2
298.51
MFCD00002754
ISYWECDDZWTKFF-UHFFFAOYSA-N
n-nonadecanoic acid, nonadecylic acid, unii-h6m3vyc62p, h6m3vyc62p, fatty acids, c16-22, n-nonadecanoate, n-nonadecylate, nonadecyclic acid, c16-c22 alkylcarboxylic acid
12591
CHEBI:39246
nonadecanoic acid
CCCCCCCCCCCCCCCCCCC(=O)O
646-30-0
C19H38O2
298.51
MFCD00002754
ISYWECDDZWTKFF-UHFFFAOYSA-N
n-nonadecanoic acid, nonadecylic acid, unii-h6m3vyc62p, h6m3vyc62p, fatty acids, c16-22, n-nonadecanoate, n-nonadecylate, nonadecyclic acid, c16-c22 alkylcarboxylic acid
12591
CHEBI:39246
nonadecanoic acid
CCCCCCCCCCCCCCCCCCC(=O)O
Boiling Point 297°C (100.0mmHg)
Melting Point 68°C to 70°C
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color White
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula CH3(CH2)17CO2H
Beilstein 02, II, 368
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot alcohol and ether
Formula Weight 298.51
Physical Form Fine Powder or Shiny Flakes
Percent Purity 98+%
Chemical Name or Material Nonadecanoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-472-4