Learn More
Leica Microsystems 3P NONCLP XTR 25X75MM SLD WHT10/C DFS Item
Manufacturer:  Leica Microsystems 3P 38002002
X‐tra Slides have a positive charged surface which helps to bond tissue sections and cytology preparations. Xtra slides are excellent for demanding H&E procedures that require tissue adhesion. Size: 1 in. x 3 in. x .04 in. (unless otherwise noted)
Catalog No. NC0044402