Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

W.A. Hammond Drierite™ Drying Desiccants

Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Mesh Size:
Nonindicating; 10-20 Mesh; 1 lb.
Nonindicating; 10-20 Mesh; 5 lb.
Nonindicating; 4 mesh; 1lb
Nonindicating; 4 Mesh; 5 lb.
Nonindicating; 6 Mesh, 1 lb.
Nonindicating; 6 Mesh; 5 lb.
Nonindicating; 8 Mesh; 1 lb.
Nonindicating; 8 mesh; 25 lb.
Nonindicating; 8 Mesh; 5 lb.
7778-18-9
CaO4S
136.134
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Abosrbent,Desiccant
24497
CHEBI:31346
calcium;sulfate
[O-]S(=O)(=O)[O-].[Ca+2]
7778-18-9
CaO4S
136.134
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Abosrbent,Desiccant
24497
CHEBI:31346
calcium;sulfate
[O-]S(=O)(=O)[O-].[Ca+2]

Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.

Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.

  • All purpose desiccant made of anhydrous calcium sulfate
  • Dries air and gases to the lowest residual moisture level attainable with a safe, easy to use desiccant (-100°F dew point)
  • Dries organic liquids to low ppm levels
  • Commonly used in desiccators, desiccator cabinets, in-line air and gas; dryers, glove boxes, sealed chambers
  • Useful for water removal from liquid reaction mixtures and multi-component;gas streams
  • Neutral, stable, constant in volume, and inert except toward water
  • Insoluble in organic solvents
  • Absorbs moisture at temperatures up to 66°C
Quantity 1 lb.
Mesh Size Nonindicating; 10-20 Mesh; 1 lb.
Packaging Glass Bottle
Chemical Name or Material Calcium Sulfate