Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm Available on GSA/VA Contract for Federal Government customers only.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.

Manufacturer: Thermo Scientific™ 906262

 View more versions of this product

Catalog No. 21-403-427

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    For Use With (Equipment) Freedom Evo™ Te-MO™ and Aquarius™
    Volume (Metric) 50μL,50µL
    Product Line Tecan
    Sterility Nonsterile
    Packaging 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case,96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
    Filtered No
    Color Clear
    Sterile No
    Features Standard
    Automated Workstation Model Freedom Evo™ Te-MO™ and Aquarius™

    Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO.

    • Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
    • Available in filtered, low retention or extended length for increased volume to reach into deep well plates
    • Lot-tested during production on actual workstation for which they are created
    • Fully conductive providing increased pipetting performance
    • Low percentage coefficient of variation (%CV)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit