Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head Available on GSA/VA Contract for Federal Government customers only.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Manufacturer: Thermo Scientific™ 919262

 View more versions of this product

Catalog No. 21-402-195

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Format 96 Tips/Rack
    For Use With (Equipment) Beckman Biomek FX/NX and 3000 and Multimek AP96 Workstations
    Color Clear
    Volume (Metric) 250μL,250µL
    Product Line Beckman
    Filtered No
    Sterility Nonsterile
    Packaging 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case,96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
    Description 919 Series, Clear
    Sterile No
    Item Description 919-262
    Tip Format Standard
    Rack Format 96 Tips/Rack
    Automated Workstation Brand Beckman Biomek FX/NX and 3000 and Multimek AP96 Workstations

    Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.  Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Beckman Biomek FX, Biomek NX, Biomek 3000 and Multimek AP96.

    • Available in low retention and extended length
    • Lot-tested during production on actual workstation for which they are created
    • Extended length tips compatible with 96-, 384- and 1536- deep well plates
    • Low percentage coefficient of variation (%CV)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit