Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonyl aldehyde, 95%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
124-19-6
C9H18O
142.24
MFCD00007030
GYHFUZHODSMOHU-UHFFFAOYSA-N
pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde
31289
CHEBI:84268
nonanal
CCCCCCCCC=O
124-19-6
C9H18O
142.24
MFCD00007030
GYHFUZHODSMOHU-UHFFFAOYSA-N
pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde
31289
CHEBI:84268
nonanal
CCCCCCCCC=O
Specific Gravity 0.827
Quantity 500mL
Flash Point 63°C
Boiling Point 93.0°C (23.0 mmHg)
Infrared Spectrum Authentic
Density 0.8270g/mL
Refractive Index 1.4230 to 1.4270
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula CH3(CH2)7CHO
Beilstein 01,708
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol
Formula Weight 142.24
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Nonyl aldehyde, 95%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 204-688-5

RTECSNumber : RA5700000

TSCA : TSCA