Learn More
Nor-Lake Scientific Enzyme Laboratory Freezers
Energy efficiency in a 24 cu. ft. freezer with heavy-duty refrigeration system
Specifications
Certifications/Compliance | CE, CEC, cUL, UL |
---|---|
Color | White |
Amperage | 15 A |
Defrost | Manual |
Dimensions (D x W x H) Interior | 30 x 23.5 x 59 in. (76 x 59 x 149 cm) |
Requires: Cord and plug for 230V 50Hz model.
Catalog Number | Mfr. No. | Frequency | Voltage | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frequency | Voltage | Price | Quantity & Availability | ||||
22-650-634
|
Nor-Lake Scientific
NSZF241WMW/0M |
60 Hz | 115 V |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
22-650-677
|
Nor-Lake Scientific
NSZF241WMW/5M |
50 Hz | 230 V |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Digital LED display microprocessor temperature controller
- Remote alarm contacts, and warm and cold audible and visual alarms
- Adjustable control range from -10°C to -25°C; factory preset at -20°C
- Continuous product temperature display
- Product sensor with glycerin bottle
- Power supply switch for manual defrost
- Air-cooled condensing unit
- Centered key-lockable doors (two keys)
- 1 in. dia. sensor access port
Construction
- Top-mounted refrigeration system
- Full-length pull door handle
- Heavy-duty solid door with spring-loaded hinges and magnetic door gasket
- Perimeter anti-condensate door frame heater wires
- Interior cabinet liner and interior door panel are aluminum
- Exterior cabinet front, sides and back painted white; top and bottom are galvanized steel
- Cabinet and solid door are foamed in-place with CFC-free high-density polyurethane foam insulation
- Four casters (two locking)
- Nine epoxy-coated wire shelves
Order Info
Optional features, including extra shelves, drawers, sensor access port, legs, stainless-steel interior/exterior, reverse door hinge, storage bins, temperature chart recorder, and 4-20ma output, available. Contact your Fisher Scientific representative for more information.
Warranty and Services
18 months parts and labor, 5 years compressor (US and Canada); 18 months parts (International)
Specifications
CE, CEC, cUL, UL | |
15 A | |
30 x 23.5 x 59 in. (76 x 59 x 149 cm) | |
R-404A | |
Upright | |
Microprocessor | |
190.05 kg |
White | |
Manual | |
34.87 x 28.5 x 79.6 in. (88 x 72 x 202 cm) | |
9 | |
-20°C | |
419 lb. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.