Nor-Lake™ Scientific Enzyme Laboratory Freezers

Energy efficiency in a 24 cu. ft. freezer with heavy-duty refrigeration system

Manufacturer: Nor-Lake Scientific NSZF241WMW/0M

 View more versions of this product

Catalog No. 22-650-634

  Check Availability
Add to Cart

Required Accessories : Cord and plug for 230V 50Hz model.

    Specifications

    Product Type Upright
    Voltage 115V
    Width (English) Exterior 28.5 in.
    Width (English) Interior 23.5 in.
    Width (Metric) Exterior 72cm
    Width (Metric) Interior 59cm
    Certifications/Compliance UL, cUL, CE, CEC
    Electrical Requirements 115V 60Hz
    Color White
    Weight (English) 419 lb.
    Weight (Metric) 190.05kg
    Temperature Sensor Microprocessor
    Amperage 15A
    Capacity 24 cu. ft., 680L
    Capacity (English) 24 cu. ft.
    Capacity (Metric) 680L
    Defrost Manual
    Depth (English) Interior 30 in.
    Depth (Metric) Interior 76cm
    Door Count 1
    Dimensions (L x W x H) Exterior 34.87 x 28.5 x 79.6 in. (88 x 72 x 202cm)
    Dimensions (D x W x H) Interior 30 x 23.5 x 59 in. (76 x 59 x 149cm)
    Height (English) Interior 59 in.
    Height (English) Exterior 79.6 in.
    Height (Metric) Exterior 202cm
    Height (Metric) Interior 149cm
    Hertz 60Hz
    Length (English) Exterior 34.87 in.
    Length (Metric) Exterior 88cm
    Shelves 9
    Refrigerant R-404A
    Temperature Range (Metric) −20°C

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit