Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Noradamantane-3-carbonyl chloride, 97%

Click to view available options
Quantity:
250mg
1g
5g
67064-09-9
C10H13ClO
184.663
MFCD09265122
FEJBSNLESASJKD-UHFFFAOYSA-N
tricyclo 3.3.1.0 3 ,? nonane-3-carbonyl chloride,hexahydro-2,5-methanopentalene-3a 1h-carbonyl chloride,3-noradamantanecarboxylic acid chloride,hexahydro-2,5-methano-pentalene-3a-carbonyl chloride,2,5-methanopentalene-3a 1h-carbonyl chloride, hexahydro,2,5-methanopentalene-3a 1h-carbonyl chloride, hexahydro-9ci
13182402
C1C2CC3CC1CC3(C2)C(=O)Cl
67064-09-9
C10H13ClO
184.663
MFCD09265122
FEJBSNLESASJKD-UHFFFAOYSA-N
tricyclo 3.3.1.0 3 ,? nonane-3-carbonyl chloride,hexahydro-2,5-methanopentalene-3a 1h-carbonyl chloride,3-noradamantanecarboxylic acid chloride,hexahydro-2,5-methano-pentalene-3a-carbonyl chloride,2,5-methanopentalene-3a 1h-carbonyl chloride, hexahydro,2,5-methanopentalene-3a 1h-carbonyl chloride, hexahydro-9ci
13182402
C1C2CC3CC1CC3(C2)C(=O)Cl
Density 1.165g/mL
Refractive Index 1.5160
Quantity 1g
Color White
Sensitivity Moisture Sensitive
UN Number UN3261
Formula Weight 184.66
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Noradamantane-3-carbonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.