Learn More
MP Biomedical™ Noradrenaline Fast Track ELISA Kit
Quantitative Enzyme-linked Immunosorbent Assay
Manufacturer:  MP Biomedical™ 07LE6200
Catalog No. MP07LE6200
Add to cart
Description
Description
The Noradrenaline Fast Track ELISA Kit is intended for the measurement of Noradrenaline (norepinephrine) in plasma and urine. Samples are extracted by using a cis-diol-specific affinity gel, acylated and then derivatized enzymatically.
Specifications
Specifications
Kit and Assay
FDA Exempt
2°C to 8°C
Plasma, Urine
Biogenic Amines
10 μL for Urin; 300 μL for Plasma
Human