Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norcamphor, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
497-38-1
C7H10O
110.16
MFCD00074823
KPMKEVXVVHNIEY-UHFFFAOYSA-N
norcamphor,2-norbornanone,bicyclo 2.2.1 heptan-2-one,norcampher,2-oxonorbornane,norbornan-2-one,2,5-methanocyclohexanone,8,9,10-trinorbornan-2-one,bicyclo 2.2.1 heptan-3-one
10345
bicyclo[2.2.1]heptan-3-one
C1CC2CC1CC2=O
497-38-1
C7H10O
110.16
MFCD00074823
KPMKEVXVVHNIEY-UHFFFAOYSA-N
norcamphor,2-norbornanone,bicyclo 2.2.1 heptan-2-one,norcampher,2-oxonorbornane,norbornan-2-one,2,5-methanocyclohexanone,8,9,10-trinorbornan-2-one,bicyclo 2.2.1 heptan-3-one
10345
bicyclo[2.2.1]heptan-3-one
C1CC2CC1CC2=O
Boiling Point 168°C to 172°C
Melting Point 88°C to 91°C
Flash Point 33°C
Quantity 100g
Color Colorless to White
Packaging Plastic bottle
Infrared Spectrum Authentic
Density 33g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 07, 57
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 110.16
Physical Form Adhering Crystals
Percent Purity ≥98.5%
Chemical Name or Material Norcamphor
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning