Norcross Safety PVC Pull-On Boots

Manufacturer: Honeywell Safety Products 18801BLM090

 View more versions of this product

Catalog No. 19-051814

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-11 M I/75 C/75
    For Use With (Application) General industrial, farming, general occupation
    Height (English) 15 in.
    Resistant To degradation from agricultural chemicals, fertilizers and animal waste
    Seam Type Seamless
    Shank Material Steel
    Sole Features Deep angle cleated sole
    Toe Material Steel
    Waterproof Yes
    Color Black
    Material PVC
    Size 9

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit