NorLake™ Scientific Select Series Temperature Stability Test Chambers w/Glass Doors
Advanced engineered design for optimal storage temperature environments
Manufacturer: Nor-Lake Scientific NSRI331WSG/0
 View more versions of this product
Catalog No. 22-650-831
  Check Availability
Add to Cart
Specifications
33.1 cu.ft.
87.62 in.
115V
31.75 in.
67 in.
31 in.
27.75 in.
14.7A
940L (33.1 cu.ft.)
LED
NEMA: 5-20P
199kg
940L
224cm
60Hz
80cm
170cm
78cm
70cm
UL, C-UL
115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R
1
R-134a
473 lb.
  • Description
  • Specifications
  • Related Products
    Product Content Feedback