Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Chicken IgY Antibody, purified / AC146 Chicken, Polyclonal, Isotype Control, MilliporeSigma™

Chicken Isotype Control

Manufacturer:  MilliporeSigma AC146

Catalog No. AC146MI


Add to cart

Description

Description

Chickens transfer high quantities of IgY (IgG) into the egg yolk and harvesting antibodies from eggs eliminates the need for the invasive bleeding procedure. One week ft.s eggs can contain 10 times more antibodies than the volume of rabbit blood obtained from one weekly bleeding. However, there are some disadvantages when using certain chicken derived antibodies in immunoassays. Chicken IgY does not fix mammalian complement component C1 and it does not perform as a precipitating antibody using standard solutions.
Specifications

Specifications

IgY
Unconjugated
Purified
IgY
100μg
Primary
1mg/mL
Purified chicken polyclonal in buffer containing PBS with 0.05% sodium azide.
Chicken
T-gel
Stable for 1 year at 2-8°C from date of receipt.
Polyclonal
Documents
Promotions

Promotions

  • MilliporeSigma
    Get 15% off MilliporeSigma antibodies with promo code 15offAB.

    12-31-2019

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only