Learn More
US Biomax Inc High density lung tissue array, 96 cases/192 cores  DFS Item
Manufacturer:  US Biomax Inc LC2086A
High density lung tissue microarray, containing 29 cases of normal lung tissue and 67 adjacent normal lung tissue, duplicate core per case.
Catalog No. NC1776651