Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Sheep IgG, Isotype Control, R&D Systems™
SDP

Catalog No. 5001A
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5001A 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5001A Supplier R&D Systems Supplier No. 5001A

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Control.

Specifications

Antigen IgG
Applications Control
Classification Polyclonal
Conjugate Unconjugated
Dilution Control
Formulation 0.05% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Sheep
Quantity 1 mg
Regulatory Status RUO
Gene ID (Entrez) 3500
Reconstitution Reconstitution default not known
Target Species Sheep
Content And Storage Store the unopened product at 2 - 8degrees C.
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.